Learn More
Amisulpride-d5, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Amisulpride-d5 |
| CAS | 1216626-17-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C17 D5 H22 N3 O4 S |
| Quantity | 1 mg |
| Synonym | 4-Amino-N-[(1-ethyl-2-pyrrolidinyl)methyl]-5-(ethylsulfonyl)-2-methoxybenzamide-d5, Aminosultopride-d5, DAN-2163-d5, Socian-d5, Solian-d5, Sulamid-d5 |
| InChI Formula | InChI=1S/C17H27N3O4S/c1-4-20-8-6-7-12(20)11-19-17(21)13-9-16(25(22,23)5-2)14(18)10-15(13)24-3/h9-10,12H,4-8,11,18H2,1-3H3,(H,19,21)/i1D3,4D2 |
| SMILES | [2H]C([2H])([2H])C([2H])([2H])N1CCCC1CNC(=O)c2cc(c(N)cc2OC)S(=O)(=O)CC |
| IUPAC Name | 4-amino-5-ethylsulfonyl-2-methoxy-N-[[1-(1,1,2,2,2-pentadeuterioethyl)pyrrolidin-2-yl]methyl]benzamide |
| Molecular Weight (g/mol) | 374.51 |
| Show More |