missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amprenavir-d4, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Amprenavir-d4 |
| CAS | 1217661-20-5 |
| Molecular Formula | C25H31D4N3O6S |
| Quantity | 1 mg |
| Synonym | 141W94-d4, KVX-478-d4, Agenerase-d4, Prozei-d4 |
| InChI Formula | InChI=1S/C25H35N3O6S/c1-18(2)15-28(35(31,32)22-10-8-20(26)9-11-22)16-24(29)23(14-19-6-4-3-5-7-19)27-25(30)34-21-12-13-33-17-21/h3-11,18,21,23-24,29H,12-17,26H2,1-2H3,(H,27,30)/t21-,23-,24+/m0/s1/i13D2,17D2 |
| SMILES | O=C(N[C@H]([C@@H](CN(S(C1=CC=C(N)C=C1)(=O)=O)CC(C)C)O)CC2=CC=CC=C2)O[C@H]3CC([2H])([2H])OC3([2H])[2H] |
| IUPAC Name | [(3S)-2,2,5,5-tetradeuteriooxolan-3-yl] N-[(2S,3R)-4-[(4-aminophenyl)sulfonyl-(2-methylpropyl)amino]-3-hydroxy-1-phenylbutan-2-yl]carbamate |
| Molecular Weight (g/mol) | 509.65 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction