missing translation for 'onlineSavingsMsg'
Learn More

Aprepitant-d4, TRC

Product Code. 30333946
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30333946

Brand: Toronto Research Chemicals TRCA7298031MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aprepitant-d4
CAS 1133387-60-6
Purity Grade Notes HPLC
Molecular Formula C23H17D4F7N4O3
Quantity 1 mg
Synonym 5-[[(2R,3S)-2-[(1R)-1-[3,5-Bis(trifluoromethyl)phenyl]ethoxy]-3-(4-fluorophenyl-d4)-4-morpholinyl]methyl]-1,2-dihydro-3H-1,2,4-triazol-3-one, Emend-d4, L 754030-d4, MK 0869-d4, MK 869-d4, ONO 7436-d4
InChI Formula InChI=1S/C23H21F7N4O3/c1-12(14-8-15(22(25,26)27)10-16(9-14)23(28,29)30)37-20-19(13-2-4-17(24)5-3-13)34(6-7-36-20)11-18-31-21(35)33-32-18/h2-5,8-10,12,19-20H,6-7,11H2,1H3,(H2,31,32,33,35)/t12-,19+,20-/m1/s1/i2D,3D,4D,5D
SMILES O=C1NC(CN2CCO[C@H](O[C@H](C)C3=CC(C(F)(F)F)=CC(C(F)(F)F)=C3)[C@@H]2C4=C([2H])C([2H])=C(F)C([2H])=C4[2H])=NN1
IUPAC Name 3-[[(2R,3S)-2-[(1R)-1-[3,5-bis(trifluoromethyl)phenyl]ethoxy]-3-(2,3,5,6-tetradeuterio-4-fluorophenyl)morpholin-4-yl]methyl]-1,4-dihydro-1,2,4-triazol-5-one
Molecular Weight (g/mol) 538.45
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.