Learn More
Tocris Bioscience™ PAMP-12 (human, porcine)
GREENER_CHOICE
Product Code. 18758649
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18758649
Brand: Tocris Bioscience™ 6551/1
Potent MRGPRX2 agonist
PAMP-12 (human, porcine) is a potent endogenous peptide agonist of Mas related GPR X2 (MRGPRX2, EC50 = 57.2 nM). Corresponds to amino acids 9 to 20 of proadrenomedullin.
TRUSTED_SUSTAINABILITY
Specifications
CAS 196305-05-2
Content And Storage Store at -20°C
Molecular Weight (g/mol) 1618.95
Quantity 1 mg