missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acebutolol Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Description
Acebutolol Hydrochloride 1.0 mg/ml in Methanol (as free base)
Specifications
Specifications
| Chemical Name or Material | Acebutolol Hydrochloride |
| CAS | 34381-68-5 |
| Analyte or Component Names | Acebutolol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H28 N2 O4 . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Acebutolol Hydrochloride |
| InChI Formula | InChI=1 S/C18H28N2O4.ClH/c1-5-6-18(23)20-14-7-8-17(16(9-14)13(4)21)24-11-15(22)10-19-12(2)3;/h7-9,12,15,19,22 H,5-6,10-11H2,1-4H3,(H,20,23);1 H |
| SMILES | Cl.CCCC(=O)Nc1ccc(OCC(O)CNC(C)C)c(c1)C(=O)C |
| IUPAC Name | N-[3-acetyl-4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]butanamide;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?