missing translation for 'onlineSavingsMsg'
Learn More

Desmethyltrimipramine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30462084
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30462084

Brand: LoGiCal LGCAMP0240.0201

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Desmethyltrimipramine Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Desmethyltrimipramine Hydrochloride
CAS 10464-24-1
Analyte or Component Names Desmethyltrimipramine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H24 N2 . Cl H
Certifications/Compliance ISO 17034
Synonym (2 RS)-3-(10,11-Dihydro-5 H-dibenzo[b,f]azepin-5-yl)-N,2-dimethylpropan-1-amine Hydrochloride (Desmethyltrimipramine Hydrochloride), Trimipramine Maleate Imp. B (EP), Trimipramine Imp. B (EP), (2 RS)-3-(10,11-Dihydro-5 H-dibenzo[b,f]azepin-5-yl)-N,2-dimethylpropan-1-amine Hydrochloride, Trimipramine Maleate Imp. B (EP) as Hydrochloride, Desmethyltrimipramine Hydrochloride, Trimipramine Maleate Impurity B as Hydrochloride, Trimipramine Impurity B as Hydrochloride
InChI Formula InChI=1 S/C19H24N2.ClH/c1-15(13-20-2)14-21-18-9-5-3-7-16(18)11-12-17-8-4-6-10-19(17)21;/h3-10,15,20 H,11-14H2,1-2H3;1 H
SMILES Cl.CNCC(C)CN1c2ccccc2CCc3ccccc13
IUPAC Name 3-(5,6-Dihydrobenzo[b][1 ]benzazepin-11-yl)-N,2-dimethylpropan-1-amine Hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 316.87
Quantity 1 mL
Formula Weight 316.1706 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.