Learn More
SPEX Certiprep 1ML tau-Fluvalinate, 1,000 µg/mL Organic Standard
Brand:  SPEX Certiprep S-4161
1ML tau-Fluvalinate, 1,000 µg/mL Organic Standard
Product Code. 16310268
  • ONLINE EXCLUSIVE PRICE
    € 198.40
    € 169.55 / 1mL
    Save € 28.85
    14% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • tau-Fluvalinate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetone