missing translation for 'onlineSavingsMsg'
Learn More

1R-Hydroxy-5-keto Tiamulin, TRC

Product Code. 30395421
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395421 25 mg 25mg
30405835 2.5 mg 2.5mg
30393693 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395421 Supplier Toronto Research Chemicals Supplier No. TRCH94276525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1R-Hydroxy-5-keto Tiamulin
Analyte or Component Names 1R-Hydroxy-5-keto Tiamulin
Shipping Condition Room Temperature
Molecular Formula C28 H47 N O4 S
Synonym (3R,3aR,4R,5R,7S,9R,9aS,12S)-3-Hydroxy-4,7,9,12-tetramethyl-8-oxo-7-vinyldecahydro-4,9a-propanocyclopenta[8]annulen-5-yl 2-((2-(Diethylamino)ethyl)thio)acetate
InChI Formula InChI=1S/C28H47NO4S/c1-8-26(6)17-22(33-23(31)18-34-16-15-29(9-2)10-3)27(7)19(4)11-13-28(20(5)25(26)32)14-12-21(30)24(27)28/h8,19-22,24,30H,1,9-18H2,2-7H3/t19-,20-,21+,22+,24-,26+,27-,28+/m0/s1
Solution Type Neat
SMILES CCN(CC)CCSCC(=O)O[C@@H]1C[C@@](C)(C=C)C(=O)[C@H](C)[C@@]23CC[C@@H](O)[C@H]2[C@@]1(C)[C@@H](C)CC3
IUPAC Name [(1S,2R,4S,6R,7R,8R,9R,14S)-4-ethenyl-9-hydroxy-2,4,7,14-tetramethyl-3-oxo-6-tricyclo[5.4.3.01,8]tetradecanyl] 2-[2-(diethylamino)ethylsulfanyl]acetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 493.742
Quantity 25 mg
Formula Weight 493.323
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.