missing translation for 'onlineSavingsMsg'
Learn More

(1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Ethoxy-1-[[2'-(1-ethyl-1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Mikromol™

Product Code. 30050516
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050516

Brand: Mikromol MM0776.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Ethoxy-1-[[2'-(1-ethyl-1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-1-Ethyl Candesartan Cilexetil
CAS 914613-35-7
Analyte or Component Names 1H-1-Ethyl Candesartan Cilexetil
Shipping Condition Room Temperature
Molecular Formula C35 H38 N6 O6
Certifications/Compliance ISO 9001
Synonym 1H-Benzimidazole-7-carboxylic acid, 2-ethoxy-1-[[2'-(1-ethyl-1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 1-[[(cyclohexyloxy)carbonyl]oxy]ethyl ester, 1-[[(Cyclohexyloxy)carbonyl]oxy]ethyl 2-ethoxy-1-[[2'-(1-ethyl-1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Candesartan Cilexetil Imp. E (EP), (1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Ethoxy-1-[[2'-(1-ethyl-1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate, (1RS)-1-[[(Cyclohexyloxy)carbonyl]-oxy]ethyl 2-Ethoxy-1-[[2'-(1-ethyl-1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate, Candesartan Cilexetil Impurity E
InChI Formula InChI=1S/C35H38N6O6/c1-4-41-32(37-38-39-41)28-15-10-9-14-27(28)25-20-18-24(19-21-25)22-40-31-29(16-11-17-30(31)36-34(40)44-5-2)33(42)45-23(3)46-35(43)47-26-12-7-6-8-13-26/h9-11,14-21,23,26H,4-8,12-13,22H2,1-3H3
SMILES CCOc1nc2cccc(C(=O)OC(C)OC(=O)OC3CCCCC3)c2n1Cc4ccc(cc4)c5ccccc5c6nnnn6CC
IUPAC Name 1-cyclohexyloxycarbonyloxyethyl 2-ethoxy-3-[[4-[2-(1-ethyltetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
Molecular Weight (g/mol) 638.71
Quantity 25 mg
Formula Weight 638.2853 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.