Learn More
(1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid, Mikromol™
Description
(1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid
Specifications
Specifications
| Chemical Name or Material | (1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid |
| CAS | 2517611-89-9 |
| Analyte or Component Names | (1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C26 H32 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1RS,4SR)-7-(2-Methylpropyl)-1-[4-(2-methyl-propyl)phenyl]-1,2,3,4-tetrahydronaphthalene-1,4-dicarboxylic Acid |
| InChI Formula | InChI=1S/C26H32O4/c1-16(2)13-18-5-8-20(9-6-18)26(25(29)30)12-11-22(24(27)28)21-10-7-19(14-17(3)4)15-23(21)26/h5-10,15-17,22H,11-14H2,1-4H3,(H,27,28)(H,29,30)/t22-,26+/m0/s1 |
| SMILES | CC(C)Cc1ccc(cc1)[C@@]2(CC[C@H](C(=O)O)c3ccc(CC(C)C)cc23)C(=O)O |
| IUPAC Name | (1S,4R)-6-(2-methylpropyl)-4-[4-(2-methylpropyl)phenyl]-2,3-dihydro-1H-naphthalene-1,4-dicarboxylic acid |
| Show More |