Learn More
(1S)-1-[3-[(E)-2-(7-Chloro-2-quinolyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propan-1-ol, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(1S)-1-[3-[(E)-2-(7-Chloro-2-quinolyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propan-1-ol
Specifications
Specifications
| Chemical Name or Material | 2-[3-(S)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]phenyl-2-propanol |
| CAS | 287930-77-2 |
| Analyte or Component Names | 2-[3-(S)-[3-(2-(7-Chloro-2-quinolinyl)ethenyl)phenyl]-3-hydroxypropyl]phenyl-2-propanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H28 Cl N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (αS)-α-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-2-(1-hydroxy-1-methylethyl)benzenepropanol, (S)-1-[3-[(E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]-1-propanol, (αS)-α-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-2-(1-hydroxy-1-methylethyl)benzenepropanol, 2-[2-[3(S)-[3-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-hydroxypropyl]phenyl-2-propanol |
| InChI Formula | InChI=1S/C29H28ClNO2/c1-29(2,33)26-9-4-3-7-21(26)13-17-28(32)23-8-5-6-20(18-23)10-15-25-16-12-22-11-14-24(30)19-27(22)31-25/h3-12,14-16,18-19,28,32-33H,13,17H2,1-2H3/b15-10+/t28-/m0/s1 |
| SMILES | CC(C)(O)c1ccccc1CC[C@H](O)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2 |
| IUPAC Name | (1S)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propan-1-ol |
| Show More |