missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2R)-1-Benzyl-3-(dimethylamino)-2-methyl-1-phenylpropyl Acetate (Acetoxyphene), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(1S,2R)-1-Benzyl-3-(dimethylamino)-2-methyl-1-phenylpropyl Acetate (Acetoxyphene)
Specifications
Specifications
| Chemical Name or Material | alpha-d-2-Acetoxy-4-dimethylamino-1,2-diphenyl-3-methylbutane |
| CAS | 33318-28-4 |
| Analyte or Component Names | alpha-d-2-Acetoxy-4-dimethylamino-1,2-diphenyl-3-methylbutane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H27 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Dextropropoxyphene Hydrochloride Imp. B (EP), Dextropropoxyphene Imp. B (EP), Propoxyphene USP Related Compound B, Propoxyphene USP RC B, (1S,2R)-1-Benzyl-3-(dimethylamino)-2-methyl-1-phenylpropyl Acetate, Acetoxyphene, alpha-d-2-Acetoxy-4-dimethylamino-1,2-diphenyl-3-methylbutane, Propoxyphene Related Compound B (USP), Propoxyphene Related Compound B, Dextropropoxyphene Hydrochloride Impurity B, Dextropropoxyphene Impurity B |
| InChI Formula | InChI=1S/C21H27NO2/c1-17(16-22(3)4)21(24-18(2)23,20-13-9-6-10-14-20)15-19-11-7-5-8-12-19/h5-14,17H,15-16H2,1-4H3/t17-,21+/m1/s1 |
| SMILES | C[C@H](CN(C)C)[C@](Cc1ccccc1)(OC(=O)C)c2ccccc2 |
| IUPAC Name | [(2S,3R)-4-(dimethylamino)-3-methyl-1,2-diphenylbutan-2-yl] acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction