missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride, Mikromol™

Product Code. 30050341
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30050341 25 mg 25mg
30050194 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050341 Supplier Mikromol Supplier No. MM0317.150025
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(1S,4S)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1S,4S)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride
CAS 55056-87-6
Analyte or Component Names (1S,4S)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H19 N . Cl H
Certifications/Compliance ISO 17025
Synonym (1S,4S)-N-Methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine Hydrochloride, Sertraline Hydrochloride Imp. B (EP), Sertraline Imp. B (EP), (1S-cis)-1,2,3,4-Tetrahydro-N-methyl-4-phenyl-1-naphthalenamine hydrochloride, Sertraline Hydrochloride Impurity B as (1S,4S)-Enantiomer Hydrochloride, Sertraline Impurity B as (1S,4S)-Enantiomer Hydrochloride
InChI Formula InChI=1S/C17H19N.ClH/c1-18-17-12-11-14(13-7-3-2-4-8-13)15-9-5-6-10-16(15)17;/h2-10,14,17-18H,11-12H2,1H3;1H/t14-,17-;/m0./s1
SMILES Cl.CN[C@H]1CC[C@@H](c2ccccc2)c3ccccc13
IUPAC Name (1S,4S)-N-methyl-4-phenyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
Molecular Weight (g/mol) 273.8
Quantity 25 mg
Formula Weight 273.1284 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.