Learn More
2-(1-Piperazinyl)pyrimidine, 98+%, Thermo Scientific Chemicals
Catalog Number p-2698762
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
20980-22-7
C8H12N4
164.21
MFCD00040742
MRBFGEHILMYPTF-UHFFFAOYSA-N
2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine
88747
2-piperazin-1-ylpyrimidine
C1CN(CCN1)C2=NC=CC=N2
20980-22-7
C8H12N4
164.21
MFCD00040742
MRBFGEHILMYPTF-UHFFFAOYSA-N
2-1-piperazinyl pyrimidine, 1-2-pyrimidyl piperazine, 2-piperazin-1-yl pyrimidine, 1-2-pyrimidinyl piperazine, 2-piperazinopyrimidine, campiron, pyrimidine, 2-1-piperazinyl, campirone, kampirone, 2-piperazin-1-yl-pyrimidine
88747
2-piperazin-1-ylpyrimidine
C1CN(CCN1)C2=NC=CC=N2
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point 29°C to 36°C
Color Yellow
Density 1.0700g/mL
Boiling Point 96°C to 97°C (0.2 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5850 to 1.588
Quantity 1 g
Beilstein 25, IV, 2083
Specific Gravity 1.07
Formula Weight 164.21
Percent Purity 98+%
Physical Form Liquid After Melting
Chemical Name or Material 2-(1-Piperazinyl)pyrimidine