Learn More
2-[1-(tert-Butoxycarbonyl)piperid-4-yl]-1,3-thiazole-4-carboxylic acid, ≥95%, Thermo Scientific™
Product Code. p-3110218
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10768643 250 mg Amber Glass Bottle 250mg
10471893 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10768643 Supplier Thermo Scientific Maybridge Supplier No. CC31001CB
Add to basket
Chemical Identifiers
CAS 668484-45-5
Molecular Formula C13H19N3O4S
Molecular Weight (g/mol) 313.372
MDL Number MFCD06658980
InChI Key FMSFMUBALWPCQM-UHFFFAOYSA-N
Synonym 2-4-tert-butoxycarbonyl piperazin-1-yl thiazole-4-carboxylic acid, 2-4-tert-butoxycarbonyl piperazin-1-yl-1,3-thiazole-4-carboxylic acid
PubChem CID 59496772
IUPAC Name 2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]-1,3-thiazole-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CS2)C(=O)O
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 312.39
Percent Purity 95%
Chemical Name or Material 2-[1-(Tert-butoxycarbonyl)-4-piperidinyl]-1, 3-thiazole-4-carboxylic acid