missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-Triazol-1-yl)acetic Acid-D4, TRC

Product Code. 30449923
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30449923

Brand: Toronto Research Chemicals TRCT898596100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(1H-1,2,4-Triazol-1-yl)acetic Acid-D4
Molecular Formula C4 D4 H N3 O2
Quantity 100 mg
Synonym 2-(1H-1,2,4-Triazol-1-yl-D2)acetic-2,2-D2 Acid
InChI Formula InChI=1S/C4H5N3O2/c8-4(9)1-7-3-5-2-6-7/h2-3H,1H2,(H,8,9)/i1D2,2D,3D
SMILES [2H]c1nc([2H])n(n1)C([2H])([2H])C(=O)O
IUPAC Name 2,2-dideuterio-2-(3,5-dideuterio-1,2,4-triazol-1-yl)acetic acid
Molecular Weight (g/mol) 131.126
Formula Weight 131.063
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.