Learn More
2-(1H-Imidazol-1-yl)acetic Acid, Mikromol™
Product Code. 17923376
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17923376 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17923376 Supplier Mikromol Supplier No. MM1453.010025
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(1H-Imidazol-1-yl)acetic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Imidazol-1-yl-acetic Acid
CAS 22884-10-2
Analyte or Component Names Imidazol-1-yl-acetic Acid
Shipping Condition Room Temperature
Molecular Formula C5 H6 N2 O2
Certifications/Compliance ISO 9001
Synonym 2-(1H-Imidazol-1-yl)acetic Acid, Imidazole-1-acetic acid (6CI,7CI,8CI), (1H-Imidazol-1-yl)acetic acid, 2-(1H-Imidazol-1-yl)acetic acid, Imidazol-1-ylacetic acid, Zoledronic Acid Related Compound A, Zoledronic Acid Impurity D
InChI Formula InChI=1S/C5H6N2O2/c8-5(9)3-7-2-1-6-4-7/h1-2,4H,3H2,(H,8,9)
SMILES OC(=O)Cn1ccnc1
IUPAC Name 2-imidazol-1-ylacetic acid
Molecular Weight (g/mol) 126.11
Quantity 25 mg
Formula Weight 126.0429 g/mol