missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethoxy]ethanol, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
2-[2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethoxy]ethanol
Specifications
Specifications
| Chemical Name or Material | 2-[2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethoxy]ethanol |
| CAS | 19765-12-9 |
| Analyte or Component Names | 2-[2-(2-Methyl-4-nitro-1H-imidazol-1-yl)ethoxy]ethanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H13 N3 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Ethanol, 2-[2-(2-methyl-4-nitro-1H-imidazol-1-yl)ethoxy]-, Ethanol, 2-[2-(2-methyl-4-nitroimidazol-1-yl)ethoxy]- (8CI) |
| InChI Formula | InChI=1S/C8H13N3O4/c1-7-9-8(11(13)14)6-10(7)2-4-15-5-3-12/h6,12H,2-5H2,1H3 |
| SMILES | Cc1nc(cn1CCOCCO)[N+](=O)[O-] |
| IUPAC Name | 2-[2-(2-methyl-4-nitroimidazol-1-yl)ethoxy]ethanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction