Learn More
2,2,2-Trifluoroethanol, TRC
Product Code. 30449943
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30449943 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30449943 Supplier Toronto Research Chemicals Supplier No. TRCT7901755G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2,2-Trifluoroethanol
CAS 75-89-8
Molecular Formula C2 H3 F3 O
Quantity 5 g
Synonym Ethanol','2,2,2-trifluoro- (6CI','8CI','9CI','ACI), 2,2,2-Trifluoroethanol (ACI), 2,2,2-Trifluoroethan-1-ol, 2,2,2-Trifluoroethyl alcohol, MeSH ID: D014270, NSC 451, TFE, β,β,β-Trifluoroethyl alcohol
InChI Formula InChI=1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2
SMILES OCC(F)(F)F
IUPAC Name 2,2,2-trifluoroethanol
Molecular Weight (g/mol) 100.04
Formula Weight 100.0136