Learn More
2-[(2,4-Dimethylphenyl)sulfanyl]aniline, TRC
Product Code. 30369289
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369289 5 g
30387741 1 g
30376476 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30369289 Supplier Toronto Research Chemicals Supplier No. TRCD4473355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1019453-85-0
Molecular Formula C14H15NS
Molecular Weight (g/mol) 229.34
Synonym 2-[(2,4-Dimethylphenyl)thio]benzenamine
IUPAC Name 2-(2,4-dimethylphenyl)sulfanylaniline
SMILES Cc1ccc(Sc2ccccc2N)c(C)c1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C14H15NS/c1-10-7-8-13(11(2)9-10)16-14-6-4-3-5-12(14)15/h3-9H,15H2,1-2H3
Formula Weight 229.0925
Chemical Name or Material 2-[(2,4-Dimethylphenyl)sulfanyl]aniline