Learn More
JT Baker 2,2,4- Trimethylpentane BAKER ANALYZED Reagent 2.5L
Brand:  JT Baker 8715.2500
2,2,4- Trimethylpentane BAKER ANALYZED Reagent 2.5
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10199730
  • ONLINE EXCLUSIVE PRICE
    € 357.00
    € 318.84 / 2.50 L
    Save € 38.16
    10% Off
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.