missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dichloro-propanoyl Chloride, TRC

Product Code. 30428408
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428408 50 mg 50mg
30427282 500 mg -
30426019 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428408 Supplier Toronto Research Chemicals Supplier No. TRCP79521350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26073-26-7
Molecular Formula C3H3OCl3
Molecular Weight (g/mol) 161.41
Synonym Pranoprofen Impurity 30
IUPAC Name 2,2-dichloropropanoyl chloride
SMILES CC(Cl)(Cl)C(=O)Cl

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H3Cl3O/c1-3(5,6)2(4)7/h1H3
Formula Weight 159.9249
Chemical Name or Material 2,2-Dichloro-propanoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.