Learn More
2,2-Difluoro-4-pentenoic acid, 97%
Product Code. 15424867
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. 15424867
Brand: Thermo Scientific Alfa Aesar H32173.03
Request Bulk or Custom Format
CAS: 55039-89-9 | C5H6F2O2 | 136.098 g/mol
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,2-Difluoro-4-pentenoic acid is involved in the iodolactonization reaction to give the corresponding gamma- lactone, which is converted to prepare prepare 5-hydroxy-3,3-difluoropiperidine.
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55039-89-9
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD09800642
InChI Key LHOKYUDUAYXFGF-UHFFFAOYSA-N
Synonym 2,2-difluoropent-4-enoicacid, 2,2-difluoro-4-pentenoic acid, 4-pentenoic acid, 2,2-difluoro, 2,2-difluoro-pent-4-enoic acid, 10.14272/lhokyuduayxfgf-uhfffaoysa-n, doi:10.14272/lhokyuduayxfgf-uhfffaoysa-n
PubChem CID 11954656
IUPAC Name 2,2-difluoropent-4-enoic acid
SMILES C=CCC(C(=O)O)(F)F
Specifications
Boiling Point 56°C to 58°C (3 mmHg)
Quantity 1 g
UN Number UN3265
Solubility Information Immiscible with water.
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material 2,2-Difluoro-4-pentenoic acid
compliance-icons compliance-icons
Product Identifier
  • 2,2-Difluoro-4-pentenoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2,2-Difluoropent-4-enoic acid