Learn More
2,2-Difluoroacetamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,2-Difluoroacetamide |
| CAS | 359-38-6 |
| Molecular Formula | C2H3F2NO |
| Quantity | 5 g |
| Synonym | Difluoroacetamide |
| InChI Formula | InChI=1S/C2H3F2NO/c3-1(4)2(5)6/h1H,(H2,5,6) |
| SMILES | NC(=O)C(F)F |
| IUPAC Name | 2,2-difluoroacetamide |
| Molecular Weight (g/mol) | 95.05 |
| Formula Weight | 95.0183 |