Learn More
2,2'-Dipyridinyl Disulfide, TRC
Product Code. 30370080
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370080 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30370080 Supplier Toronto Research Chemicals Supplier No. TRCD4927501G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,2'-Dipyridinyl Disulfide
CAS 2127-03-9
Molecular Formula C10 H8 N2 S2
Quantity 1 g
Synonym 2,2'-Dithiobis-pyridine, 1,2-Bis(2-pyridinyl) Disulfide, 2,2'-Bipyridyldisulfide, 2,2'-Dithiobis(pyridine), 2,2'-Dithiodipyridine, 2-Aldrithiol, Di-2-pyridyl Disulfide, NSC 677438, NSC 94055
InChI Formula InChI=1S/C10H8N2S2/c1-3-7-11-9(5-1)13-14-10-6-2-4-8-12-10/h1-8H
SMILES S(Sc1ccccn1)c2ccccn2
IUPAC Name 2-(pyridin-2-yldisulfanyl)pyridine
Molecular Weight (g/mol) 220.31
Formula Weight 220.0129