Learn More
2-(2-Pyridyl)imidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2-Pyridyl)imidazole |
| CAS | 18653-75-3 |
| Molecular Formula | C8 H7 N3 |
| Quantity | 500 mg |
| Synonym | 2-(1H-Imidazol-2-yl)pyridine, 2-(2-Pyridyl)-1H-imidazole, 2-(2'-Pyridyl)imidazole, 2-(Imidazol-2-yl)pyridine, 2-(Pyridin-2-yl)imidazole, 2-(Pyridyl-2-yl)imidazole, 2-Pyridin-2-yl-1H-imidazole |
| InChI Formula | InChI=1S/C8H7N3/c1-2-4-9-7(3-1)8-10-5-6-11-8/h1-6H,(H,10,11) |
| SMILES | c1ccc(nc1)c2ncc[nH]2 |
| IUPAC Name | 2-(1H-imidazol-2-yl)pyridine |
| Molecular Weight (g/mol) | 145.161 |
| Formula Weight | 145.064 |