Learn More
2,2'-Sulfonyldiethanol, TRC
Specifications
Specifications
| Chemical Name or Material | Thiodiglycol sulfone |
| CAS | 2580-77-0 |
| Molecular Formula | C4 H10 O4 S |
| Quantity | 25 g |
| Synonym | Ethanol','2,2'-sulfonylbis-, Ethanol','2,2'-sulfonyldi- (6CI,8CI), 2,2'-Sulfonylbis[ethanol], 2,2'-Sulfonyldi[ethanol], 2-(2-Hydroxyethanesulfonyl)ethan-1-ol, 2-(2-Hydroxyethylsulfonyl)ethanol, BHES 50, Bis(2-hydroxyethyl) sulfone, Bis(β-hydroxyethyl) sulfone, NSC 32716, NSC 50845, β,β'-Dihydroxyethyl sulfone, β-Hydroxyethyl sulfone |
| InChI Formula | InChI=1S/C4H10O4S/c5-1-3-9(7,8)4-2-6/h5-6H,1-4H2 |
| SMILES | OCCS(=O)(=O)CCO |
| IUPAC Name | 2-(2-hydroxyethylsulfonyl)ethanol |
| Molecular Weight (g/mol) | 154.185 |
| Formula Weight | 154.03 |