missing translation for 'onlineSavingsMsg'
Learn More

2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, Dr. Ehrenstorfer

Product Code. 30312310
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30312310 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312310 Supplier Dr. Ehrenstorfer Supplier No. DREC10539520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol
CAS 25973-55-1
Analyte or Component Names 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol
Shipping Condition +20°C
Molecular Formula C22 H29 N3 O
Certifications/Compliance ISO 17034
Synonym Phenol, 2-(2H-benzotriazol-2-yl)-4,6-di-tert-pentyl- (7CI,8CI), 2-(2H-Benzotriazol-2-yl)-4,6-bis(1,1-dimethylpropyl)phenol, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)benzotriazole, 2-(2-Hydroxy-3,5-di-tert-pentylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, 2-(2'-Hydroxy-3',5'-di-tert-amylphenyl)benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)benzotriazole, 2-(3',5'-Di-tert-amyl-2'-hydroxyphenyl)benzotriazole, 2-(Benzotriazol-2-yl)-4,6-bis-(2-methylbutan-2-yl)phenol, 2-[2-Hydroxy-3,5-bis(1,1-dimethylpropyl)phenyl]-2H-benzotriazole, Chisorb 328, Cyasorb UV 2337, Eversorb 74, Kemisorb 74, Light Stabilizer 328, Lowilite 28, Seesorb 704, Sumisorb 350, THUV 328, Tin 328, Tinuvin 328, Tinuvin PA 238, UV 2337, UV 328, UV 74, Viosorb 591
InChI Formula InChI=1S/C22H29N3O/c1-7-21(3,4)15-13-16(22(5,6)8-2)20(26)19(14-15)25-23-17-11-9-10-12-18(17)24-25/h9-14,26H,7-8H2,1-6H3
Solution Type Neat
SMILES CCC(C)(C)c1cc(c(O)c(c1)C(C)(C)CC)n2nc3ccccc3n2
IUPAC Name 2-(benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
Industry Type Environmental Testing
Molecular Weight (g/mol) 351.49
Quantity 50 mg
Formula Weight 351.2311 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.