missing translation for 'onlineSavingsMsg'
Learn More

2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, TRC

Product Code. 30332450
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30332450 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30332450 Supplier Toronto Research Chemicals Supplier No. TRCB2070451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol
CAS 25973-55-1
Purity Grade Notes HPLC
Molecular Formula C22 H29 N3 O
Quantity 1 g
Synonym Phenol, 2-(2H-benzotriazol-2-yl)-4,6-di-tert-pentyl- (7CI,8CI), 2-(2H-Benzotriazol-2-yl)-4,6-bis(1,1-dimethylpropyl)phenol, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)benzotriazole, 2-(2-Hydroxy-3,5-di-tert-pentylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, 2-(2'-Hydroxy-3',5'-di-tert-amylphenyl)benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)benzotriazole, 2-(3',5'-Di-tert-amyl-2'-hydroxyphenyl)benzotriazole, 2-(Benzotriazol-2-yl)-4,6-bis-(2-methylbutan-2-yl)phenol, 2-[2-Hydroxy-3,5-bis(1,1-dimethylpropyl)phenyl]-2H-benzotriazole, Chisorb 328, Cyasorb UV 2337, Eversorb 74, Kemisorb 74, Light Stabilizer 328, Lowilite 28, Seesorb 704, Sumisorb 350, THUV 328, Tin 328, Tinuvin 328, Tinuvin PA 238, UV 2337, UV 328, UV 74, Viosorb 591
InChI Formula InChI=1S/C22H29N3O/c1-7-21(3,4)15-13-16(22(5,6)8-2)20(26)19(14-15)25-23-17-11-9-10-12-18(17)24-25/h9-14,26H,7-8H2,1-6H3
SMILES CCC(C)(C)c1cc(c(O)c(c1)C(C)(C)CC)n2nc3ccccc3n2
IUPAC Name 2-(benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol
Molecular Weight (g/mol) 351.49
Formula Weight 351.2311
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.