missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol |
| CAS | 25973-55-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H29 N3 O |
| Quantity | 25 g |
| Synonym | Phenol, 2-(2H-benzotriazol-2-yl)-4,6-di-tert-pentyl- (7CI,8CI), 2-(2H-Benzotriazol-2-yl)-4,6-bis(1,1-dimethylpropyl)phenol, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)-2H-benzotriazole, 2-(2-Hydroxy-3,5-di-tert-amylphenyl)benzotriazole, 2-(2-Hydroxy-3,5-di-tert-pentylphenyl)benzotriazole, 2-(2H-Benzotriazol-2-yl)-4,6-di-tert-pentylphenol, 2-(2'-Hydroxy-3',5'-di-tert-amylphenyl)benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-amyl-2-hydroxyphenyl)benzotriazole, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)-2H-benzotriazole, 2-(3,5-Di-tert-pentyl-2-hydroxyphenyl)benzotriazole, 2-(3',5'-Di-tert-amyl-2'-hydroxyphenyl)benzotriazole, 2-(Benzotriazol-2-yl)-4,6-bis-(2-methylbutan-2-yl)phenol, 2-[2-Hydroxy-3,5-bis(1,1-dimethylpropyl)phenyl]-2H-benzotriazole, Chisorb 328, Cyasorb UV 2337, Eversorb 74, Kemisorb 74, Light Stabilizer 328, Lowilite 28, Seesorb 704, Sumisorb 350, THUV 328, Tin 328, Tinuvin 328, Tinuvin PA 238, UV 2337, UV 328, UV 74, Viosorb 591 |
| InChI Formula | InChI=1S/C22H29N3O/c1-7-21(3,4)15-13-16(22(5,6)8-2)20(26)19(14-15)25-23-17-11-9-10-12-18(17)24-25/h9-14,26H,7-8H2,1-6H3 |
| SMILES | CCC(C)(C)c1cc(c(O)c(c1)C(C)(C)CC)n2nc3ccccc3n2 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4,6-bis(2-methylbutan-2-yl)phenol |
| Molecular Weight (g/mol) | 351.49 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?