missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,6-Tetrachlorotoluene, TRC

Product Code. 17933105
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933105 10 mg 10mg
17994045 25 mg 25mg
17953105 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933105 Supplier Toronto Research Chemicals Supplier No. TRCT29205510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 875-40-1
Molecular Formula C7H4Cl4
Molecular Weight (g/mol) 229.92
Synonym 2,3,4,6-Tetrachlorotoluene, 1,2,3,5-Tetrachloro-4-methylbenzene (ACI), Toluene, 2,3,4,6-tetrachloro- (6CI, 7CI, 8CI), 2,3,4,6-Tetrachlorotoluene, 1,2,3,5-tetrachloro-4-methylbenzene
IUPAC Name 1,2,3,5-tetrachloro-4-methylbenzene
SMILES Cc1c(Cl)cc(Cl)c(Cl)c1Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C7H4Cl4/c1-3-4(8)2-5(9)7(11)6(3)10/h2H,1H3
Formula Weight 227.9067
Chemical Name or Material 2,3,4,6-Tetrachlorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.