Learn More
2-(3,5-Bis(trifluoromethyl)phenyl)acetonitrile, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 85068-32-2 |
| Molecular Formula | C10H5F6N |
| MDL Number | MFCD00009904 |
| Quantity | 100 g |
| SMILES | N#CCC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1 |
| Molecular Weight (g/mol) | 253.14 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-(3,5-Bis(trifluoromethyl)phenyl)acetonitrile |