missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Trimethylpyridine, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2,3,5-Trimethylpyridine |
| CAS | 695-98-7 |
| Molecular Formula | C8H11N |
| Quantity | 5 g |
| Synonym | 2,3,5-Collidine, 2,3,5-Trimethyl-pyridine |
| InChI Formula | InChI=1S/C8H11N/c1-6-4-7(2)8(3)9-5-6/h4-5H,1-3H3 |
| SMILES | Cc1cnc(C)c(C)c1 |
| IUPAC Name | 2,3,5-trimethylpyridine |
| Molecular Weight (g/mol) | 121.18 |
| Formula Weight | 121.0891 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction