Learn More
2,3,6-Trifluoropyridine, 97%
Product Code. 11348926
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11348926
Brand: Thermo Scientific Alfa Aesar L19540.06
Request Bulk or Custom Format
CAS: 3512-18-3 | C5H2F3N | 133.073 g/mol
2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,6-Trifluoropyridine is used as an intermediate in organic syntheses and in pharmaceuticals.
Solubility
Not miscible or difficult to mix in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3512-18-3
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001158
InChI Key HRLIANGJWPJFKW-UHFFFAOYSA-N
Synonym pyridine, 2,3,6-trifluoro, 2,3,6-trifluoro-pyridine, pubchem7955, acmc-1cs48, 2,3,6-tris fluoranyl pyridine
PubChem CID 2783288
IUPAC Name 2,3,6-trifluoropyridine
SMILES C1=CC(=NC(=C1F)F)F
Specifications
Density 1.402
Boiling Point 100°C to 102°C
Flash Point 30°C (86°F)
Refractive Index 1.42
Quantity 5 g
UN Number UN1993
Beilstein 1365146
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 133.07
Percent Purity 97%
Chemical Name or Material 2,3,6-Trifluoropyridine