Learn More
2,3-Diaminopyridine, 98%
Product Code. 11497556
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11497556
Brand: Thermo Scientific Alfa Aesar A14618.14
Request Bulk or Custom Format
CAS: 452-58-4 | C5H7N3 | 109.13 g/mol
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3-Diaminopyridine was used in the synthesis of 2,3-diaminopyridinium-4-hydroxybenzoate1. It was used as internal standard in analysis of poly(amidoamine)dendrimers having different terminal functionalities by capillary electrophoresis. It was used as reagent in synthesis of imidazopyridines and organometallic complexes.
Solubility
Soluble in water (100 g/L).
Notes
It is sensitive to air. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 452-58-4
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006319
InChI Key ZZYXNRREDYWPLN-UHFFFAOYSA-N
Synonym 2,3-diaminopyridine, 2,3-pyridinediamine, pyridine, 2,3-diamino, pyridine-2,3-diyldiamine, unii-cbx394737h, diaminopyridine, 2,3-diamino-pyridine, pyridinediamine, pyridine,3-diamino, pubchem1296
PubChem CID 9956
IUPAC Name pyridine-2,3-diamine
SMILES NC1=CC=CN=C1N
Specifications
Melting Point 110°C to 114°C
Boiling Point 195°C (55 mmHg)
Flash Point 205°C (401°F)
Quantity 25 g
UN Number UN2671
Beilstein 109869
Sensitivity Air Sensitive
Solubility Information Soluble in water (100g/L).
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 2,3-Diaminopyridine