missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dihydroxypropyl Salicylate, TRC

Product Code. 30368192
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368192 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30368192 Supplier Toronto Research Chemicals Supplier No. TRCD267950100MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,3-Dihydroxypropyl Salicylate
CAS 6170-45-2
Molecular Formula C10 H12 O5
Quantity 100 mg
Synonym 2,3-Dihydroxypropyl-2-hydroxybenzoate, Glycerol α-Monosalicylate
InChI Formula InChI=1S/C10H12O5/c11-5-7(12)6-15-10(14)8-3-1-2-4-9(8)13/h1-4,7,11-13H,5-6H2
SMILES OCC(O)COC(=O)c1ccccc1O
IUPAC Name 2,3-dihydroxypropyl 2-hydroxybenzoate
Molecular Weight (g/mol) 212.199
Formula Weight 212.068
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.