Learn More
2-(4-(2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl)-2,3-dichlorophenoxy)acetic Acid, TRC
Product Code. 30350964
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350964 10 mg 10mg
30367046 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30350964 Supplier Toronto Research Chemicals Supplier No. TRCC17868010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25355-92-4
Molecular Formula C26 H24 Cl4 O8
Molecular Weight (g/mol) 606.28
Synonym Acetic acid, [4-[2-[4-(carboxymethoxy)-2,3-dichlorobenzoyl]-3,4-dihydro-2,5-diethyl-2H-pyran-6-yl]-2,3-dichlorophenoxy]- (9CI), Acetic acid, [4-[2-(α-carboxy-2,3-dichloro-p-anisoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3-dichlorophenoxy]- (8CI), 2-(4-(2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl)-2,3-dichlorophenoxy)acetic acid, [4-[2-[4-(Carboxymethoxy)-2,3-dichlorobenzoyl]-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl]-2,3-dichlorophenoxy]acetic acid
IUPAC Name 2-[4-[2-[4-(carboxymethoxy)-2,3-dichlorobenzoyl]-2,5-diethyl-3,4-dihydropyran-6-yl]-2,3-dichlorophenoxy]acetic acid
SMILES CCC1=C(OC(CC)(CC1)C(=O)c2ccc(OCC(=O)O)c(Cl)c2Cl)c3ccc(OCC(=O)O)c(Cl)c3Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C26H24Cl4O8/c1-3-13-9-10-26(4-2,25(35)15-6-8-17(23(30)21(15)28)37-12-19(33)34)38-24(13)14-5-7-16(22(29)20(14)27)36-11-18(31)32/h5-8H,3-4,9-12H2,1-2H3,(H,31,32)(H,33,34)
Formula Weight 604.0225
Chemical Name or Material 2-(4-(2-(4-(Carboxymethoxy)-2,3-dichlorobenzoyl)-2,5-diethyl-3,4-dihydro-2H-pyran-6-yl)-2,3-dichlorophenoxy)acetic Acid