missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Trichloroaniline, TRC

Product Code. 30137314
Change view
Click to view available options
Quantity:
10 g
250 g
Unit Size:
10g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137314 250 g 250g
30005779 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137314 Supplier Toronto Research Chemicals Supplier No. TRCT774090250G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 636-30-6
Molecular Formula C6 H4 Cl3 N
Molecular Weight (g/mol) 196.46
Synonym Benzenamine, 2,4,5-trichloro- (9CI, ACI), 2,4,5-Trichlorobenzenamine (ACI), Aniline, 2,4,5-trichloro- (6CI, 7CI, 8CI), 2,4,5-Trichloraniline, 2,4,5-Trichloro-phenylamine, 2,4,5-Trichloroaniline, NSC 64139
IUPAC Name 2,4,5-trichloroaniline
SMILES Nc1cc(Cl)c(Cl)cc1Cl

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H4Cl3N/c7-3-1-5(9)6(10)2-4(3)8/h1-2H,10H2
Formula Weight 194.9409
Chemical Name or Material 2,4,5-Trichloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.