Learn More
2,4,6-Trifluoropyridine, 97%
Product Code. 11368926
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11368926 1 g 1g
11378926 5 g 5g
11388926 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11368926 Supplier Thermo Scientific Alfa Aesar Supplier No. L19541.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
CAS: 3512-17-2 | C5H2F3N | 133.073 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3512-17-2
Molecular Formula C5H2F3N
Molecular Weight (g/mol) 133.073
MDL Number MFCD03001161
InChI Key UZDRWXKBKVVUTE-UHFFFAOYSA-N
Synonym 2,4,6-trifluoro-pyridine, pyridine, 2,4,6-trifluoro, pubchem7794, 2,4,6 trifluoropyridine, acmc-1cp7f, 2,4,6-tris fluoranyl pyridine
PubChem CID 2783289
IUPAC Name 2,4,6-trifluoropyridine
SMILES C1=C(C=C(N=C1F)F)F
Specifications
Boiling Point 102°C
Flash Point 17°C (62°F)
Quantity 1 g
UN Number UN2924
Beilstein 1364338
Formula Weight 133.07
Percent Purity 97%
Chemical Name or Material 2,4,6-Trifluoropyridine