Learn More
2,4'-Bipyridine, 97%
Product Code. p-7029854
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15493667 250 mg 250mg
11367139 1 g 1g
2 options
Product Code. 15493667
Brand: Thermo Scientific Alfa Aesar H27366.MD
Request Bulk or Custom Format
CAS: 581-47-5 | C10H8N2 | 156.188 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 581-47-5
Molecular Formula C10H8N2
Molecular Weight (g/mol) 156.188
MDL Number MFCD00006217
InChI Key RMHQDKYZXJVCME-UHFFFAOYSA-N
Synonym 2,4'-bipyridine, 2,4'-dipyridyl, 4-pyridylpyridine, 2,4'-bipyridyl, 2,4-bipyridyl, 2,4'-dipyridine, unii-c6sn6ru851, ccris 3428, 2-4-pyridyl pyridine, 4-pyridyl-2-pyridine
PubChem CID 68488
IUPAC Name 2-pyridin-4-ylpyridine
SMILES C1=CC=NC(=C1)C2=CC=NC=C2
Specifications
Melting Point 59°C to 63°C
Boiling Point 281°C
Quantity 250 mg
UN Number UN2811
Formula Weight 156.19
Percent Purity 97%
Chemical Name or Material 2,4'-Bipyridine