missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Bromophenyl)-5-fluoroindole, 97%

Brand:  Alfa Aesar™ H63091.03

Additional Details : CAS : 885266-74-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439588

  • € 131.00 / 1g


In Stock 1
Add to basket

Description

Description

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Chemical Identifiers

885266-74-0
290.135
XAKLXLIHCVKNII-UHFFFAOYSA-N
24720950
C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br
C14H9BrFN
MFCD05224214
2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
2-(4-bromophenyl)-5-fluoro-1H-indole

Specifications

2-(4-Bromophenyl)-5-fluoroindole
885266-74-0
MFCD05224214
XAKLXLIHCVKNII-UHFFFAOYSA-N
2-(4-bromophenyl)-5-fluoro-1H-indole
24720950
Solid
1g
C14H9BrFN
2-4-bromophenyl-5-fluoroindole, 2-4-bromophenyl-5-fluoro-1h-indole
C1=CC(=CC=C1C2=CC3=C(N2)C=CC(=C3)F)Br
290.135
290.13
97%
Videos
Documents