missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)imidazo[1,2-a]pyridine, Thermo Scientific™

Catalog Number 11470070
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
34658-66-7
C13H9BrN2
273.13
MFCD00218250
GRZUOGFRIHABDK-UHFFFAOYSA-N
2-4-bromophenyl imidazo 1,2-a pyridine, 2-4-bromo-phenyl-imidazo 1,2-a pyridine, 2-4-bromophenyl h-imidazo 1,2-a pyridine, acmc-1aihf, 2-4-bromophenyl-imidazo 1,2-a pyridine, 2-4-bromophenyl imidazo 1,2-? pyridine, 2-4-bromophenyl imidazo 1,2-a pyridine #, imidazo 1,2-a pyridine,2-4-bromophenyl, imidazol 1,2-a pyridine, 2-4-bromophenyl
623416
2-(4-bromophenyl)imidazo[1,2-a]pyridine
BrC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1
This item is not returnable. View return policy
34658-66-7
C13H9BrN2
273.13
MFCD00218250
GRZUOGFRIHABDK-UHFFFAOYSA-N
2-4-bromophenyl imidazo 1,2-a pyridine, 2-4-bromo-phenyl-imidazo 1,2-a pyridine, 2-4-bromophenyl h-imidazo 1,2-a pyridine, acmc-1aihf, 2-4-bromophenyl-imidazo 1,2-a pyridine, 2-4-bromophenyl imidazo 1,2-? pyridine, 2-4-bromophenyl imidazo 1,2-a pyridine #, imidazo 1,2-a pyridine,2-4-bromophenyl, imidazol 1,2-a pyridine, 2-4-bromophenyl
623416
2-(4-bromophenyl)imidazo[1,2-a]pyridine
BrC1=CC=C(C=C1)C1=CN2C=CC=CC2=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-(4-Bromophenyl)imidazo[1,2-a]pyridine is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.

Notes
Incompatible with oxidizing agents. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 273.13
Chemical Name or Material 2-(4-Bromophenyl)imidazo[1,2-a]pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'