missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,4-Dibromo-6-(trifluoromethyl)aniline, 97%

Product Code. 11435626
Change view
Click to view available options
Quantity:
1g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435626 1g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11435626 Supplier Alfa Aesar™ Supplier No. B20756.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 71757-14-7
Boiling Point 57°C to 59°C (0.7mmHg)
Molecular Formula C7H4Br2F3N
MDL Number MFCD00178769
Quantity 1g
UN Number UN2811
Synonym 2,4-dibromo-6-trifluoromethyl aniline, 2-amino-3,5-dibromobenzotrifluoride, benzenamine,2,4-dibromo-6-trifluoromethyl, 2,4-dibromo-6-trifluoromethyl-phenylamine, 4,6-dibromo-2-trifluoromethyl phenylamine, pubchem2759, acmc-209ojz, buttpark 29\06-98, zerenex e/9071924, 2,4-bis bromanyl-6-trifluoromethyl aniline
InChI Key ZYBKXMTWWFTYKN-UHFFFAOYSA-N
SMILES C1=C(C=C(C(=C1Br)N)C(F)(F)F)Br
IUPAC Name 2,4-dibromo-6-(trifluoromethyl)aniline
Molecular Weight (g/mol) 318.919
PubChem CID 737213
Formula Weight 318.93
Percent Purity 97%
Chemical Name or Material 2,4-Dibromo-6-(trifluoromethyl)aniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.