Learn More
2,4-Dichloro-5-methylpyrimidine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-5-methylpyrimidine**Toxic, Stench** |
| CAS | 1780-31-0 |
| Molecular Formula | C5 H4 Cl2 N2 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C5H4Cl2N2/c1-3-2-8-5(7)9-4(3)6/h2H,1H3 |
| SMILES | Cc1cnc(Cl)nc1Cl |
| IUPAC Name | 2,4-dichloro-5-methylpyrimidine |
| Molecular Weight (g/mol) | 163 |
| Formula Weight | 161.9752 |