Learn More
2,4-Dichloro-5-nitrophenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-5-nitrophenol |
| CAS | 39489-77-5 |
| Molecular Formula | C6H3Cl2NO3 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C6H3Cl2NO3/c7-3-1-4(8)6(10)2-5(3)9(11)12/h1-2,10H |
| SMILES | O=[N](C(C=C(O)C(Cl)=C1)=C1Cl)=O |
| IUPAC Name | 2,4-dichloro-5-nitrophenol |
| Molecular Weight (g/mol) | 208 |
| Formula Weight | 206.95 |