missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzotrifluoride, TRC

Product Code. 30007069
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007069 1 g 1g
30011644 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007069 Supplier Toronto Research Chemicals Supplier No. TRCD4319701G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-60-5
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 215
Synonym 1,3-Dichloro-4-(trifluoromethyl)benzene, 2,4-Dichloro(trifluoromethyl)benzene, 2,4-Dichloro-1-(trifluoromethyl)benzene, 2,4-Dichloro-α,α,α-trifluorotoluene, 2,4-Dichloro-α,α,α-trifluorotoluene, 2,4-Dichloro-1-(trifluoromethyl)benzene
SMILES FC(F)(F)c1ccc(Cl)cc1Cl

Specifications

Quantity 1 g
Chemical Name or Material 2,4-Dichlorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.