missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorobenzyl EP Impurity A, Mikromol™

Product Code. 17991186
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991186 100 mg 100mg
17970686 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991186 Supplier Mikromol Supplier No. MM0739.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2,4-Dichlorobenzyl EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2,5-dichlorophenyl)methanol
CAS 34145-05-6
Analyte or Component Names (2,5-dichlorophenyl)methanol
Shipping Condition Room Temperature
Molecular Formula C7 H6 Cl2 O
Certifications/Compliance ISO 9001
Synonym (2,5-Dichlorophenyl)methanol, 2,5-Dichlorobenzenemethanol, 2,5-Dichlorobenzyl alcohol, 2,4-Dichlorobenzyl alcohol Imp. A (EP), Dichlorobenzyl Alcohol Imp. A (EP), 2,4-Dichlorobenzyl Alcohol Impurity A
InChI Formula InChI=1S/C7H6Cl2O/c8-6-1-2-7(9)5(3-6)4-10/h1-3,10H,4H2
SMILES OCc1cc(Cl)ccc1Cl
IUPAC Name (2,5-dichlorophenyl)methanol
Molecular Weight (g/mol) 177.03
Quantity 100 mg
Formula Weight 175.9796 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.