missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxybenzaldehyde, 97%, Thermo Scientific™

Product Code. 11426070
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11426070

Brand: Thermo Scientific Alfa Aesar H26597.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Difluoro-3-methoxybenzaldehyde is used as a laboratory reagent. It is also used as an intermediate in pharmaceutical industries.

Solubility
Insoluble in water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870837-66-4
Molecular Formula C8H6F2O2
Molecular Weight (g/mol) 172.13
MDL Number MFCD04115930
InChI Key CEEXKEWRNDNTKK-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxy-benzaldehyde, benzaldehyde,2,4-difluoro-3-methoxy, 2,4-bis fluoranyl-3-methoxy-benzaldehyde
PubChem CID 3720237
IUPAC Name 2,4-difluoro-3-methoxybenzaldehyde
SMILES COC1=C(F)C=CC(C=O)=C1F

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 2,4-Difluoro-3-methoxybenzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.