Learn More
(2,4-Difluoro-6-methoxyphenyl)boronic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | (2,4-Difluoro-6-methoxyphenyl)boronic Acid |
| CAS | 1808997-86-5 |
| Molecular Formula | C7H7BF2O3 |
| Quantity | 500 mg |
| Synonym | B-(2,4-Difluoro-6-methoxyphenyl)boronic acid |
| InChI Formula | InChI=1S/C7H7BF2O3/c1-13-6-3-4(9)2-5(10)7(6)8(11)12/h2-3,11-12H,1H3 |
| SMILES | COc1cc(F)cc(F)c1B(O)O |
| IUPAC Name | (2,4-difluoro-6-methoxyphenyl)boronic acid |
| Molecular Weight (g/mol) | 187.94 |
| Formula Weight | 188.0456 |