Learn More
2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals
Catalog Number p-3736241
Click to view available options
Quantity:
5 g
25 g
626-03-9
C5H5NO2
111.1
MFCD00006273
ZEZJPIDPVXJEME-UHFFFAOYSA-N
2,4-dihydroxypyridine, pyridine-2,4-diol, 2,4-pyridinediol, 4-hydroxypyridin-2 1h-one, 3-deazauracil, 4-hydroxy-2-pyridone, 2 1h-pyridinone, 4-hydroxy, 2,4-dihydroxy pyridine, 2-hydroxy-1h-pyridin-4-one, 4-hydroxy-2 1h-pyridinone
54696004
4-hydroxy-1H-pyridin-2-one
C1=CNC(=O)C=C1O
626-03-9
C5H5NO2
111.1
MFCD00006273
ZEZJPIDPVXJEME-UHFFFAOYSA-N
2,4-dihydroxypyridine, pyridine-2,4-diol, 2,4-pyridinediol, 4-hydroxypyridin-2 1h-one, 3-deazauracil, 4-hydroxy-2-pyridone, 2 1h-pyridinone, 4-hydroxy, 2,4-dihydroxy pyridine, 2-hydroxy-1h-pyridin-4-one, 4-hydroxy-2 1h-pyridinone
54696004
4-hydroxy-1H-pyridin-2-one
C1=CNC(=O)C=C1O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 278.0°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H3N)(OH)2
Quantity 5 g
Beilstein 21, 160
Formula Weight 111.1
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 4-Dihydroxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dihydroxypyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.